Learn More
Medchemexpress LLC HY-10219 10mg , Rapamycin CAS:53123-88-9 Purity:>98%
SDP
Catalog No. 502258958
Encompass_Preferred
HY-10219 10mg Rapamycin CAS: 53123-88-9 Rapamycin (Sirolimus; AY 22989) is a potent and specific mTOR inhibitor with an IC50 of 0.1 nM in HEK293 cells. Rapamycin binds to FKBP12 and specifically acts as an allosteric inhibitor of mTORC1. Rapamycin is an autophagy activator, an immunosuppressant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-8958 Supplier Medchemexpress LLC Supplier No. HY1021910MG
Add to Cart
Add to Cart
HY-10219 10mg Rapamycin CAS: 53123-88-9 Rapamycin (Sirolimus; AY 22989) is a potent and specific mTOR inhibitor with an IC50 of 0.1 nM in HEK293 cells. Rapamycin binds to FKBP12 and specifically acts as an allosteric inhibitor of mTORC1. Rapamycin is an autophagy activator, an immunosuppressant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.