Learn More
Medchemexpress LLC Rapamycin-13C,d3 (sirolimus-13C,d3) | 00-00-0 | MFCD00867594 | 97.1% | 918.18 g/mol | C50 13CH76D3NO13 | 500 UG
SDP
Catalog No. 5000246752
Encompass_Preferred
Rapamycin-13C,d3 is a 13C- and deuterium-labeled isotopologue of rapamycin supplied for use as an analytical/internal standard and for tracer studies. The product is provided as a 500 μg quantity with a reported purity of ~97%, and is intended for research use with accompanying product information and safety documentation.
  • Isotopically labeled with 13C and d3 for clear mass-shift detection.
  • Supplied as a 500 μg quantity suitable for analytical assays.
  • Reported purity approximately 97.05% for reliable quantitation.
  • Suitable as an internal standard for LC-MS/MS and tracer studies.
  • Product information and MSDS available for handling and storage guidance.
Catalog No. 50-002-46752 Supplier Medchemexpress LLC Supplier No. HY10219S1500UG
Rapamycin-13C,d3 is a 13C- and deuterium-labeled isotopologue of rapamycin supplied for use as an analytical/internal standard and for tracer studies. The product is provided as a 500 μg quantity with a reported purity of ~97%, and is intended for research use with accompanying product information and safety documentation.
  • Isotopically labeled with 13C and d3 for clear mass-shift detection.
  • Supplied as a 500 μg quantity suitable for analytical assays.
  • Reported purity approximately 97.05% for reliable quantitation.
  • Suitable as an internal standard for LC-MS/MS and tracer studies.
  • Product information and MSDS available for handling and storage guidance.