Learn More
Medchemexpress LLC HY-10219 1g , Rapamycin CAS:53123-88-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY102191G
Encompass_Preferred
HY-10219 1g Rapamycin CAS: 53123-88-9 Rapamycin (Sirolimus; AY 22989) is a potent and specific mTOR inhibitor with an IC50 of 0.1 nM in HEK293 cells. Rapamycin binds to FKBP12 and specifically acts as an allosteric inhibitor of mTORC1. Rapamycin is an autophagy activator, an immunosuppressant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9293