Learn More
BIOGEMS INTERNATIONAL INC Rapamycin 1mg
SDP
Catalog No. 501530184
Encompass
Rapamycin was first evaluated as an antifungal agent, but is now studied for its potent immunosuppressive and antiproliferative properties. It forms a complex with FKBP12 and directly binds to mTORC1, inhibiting the functions of the mTOR pathway. Rapamycin is reported to block protein synthesis, arrest cell cycle progression, and inhibit IL-2 signal transduction mechanisms.
Catalog No. 50-153-0184 Supplier BIOGEMS INTERNATIONAL INC Supplier No. 53188931MG
Add to Cart
Add to Cart
Rapamycin was first evaluated as an antifungal agent, but is now studied for its potent immunosuppressive and antiproliferative properties. It forms a complex with FKBP12 and directly binds to mTORC1, inhibiting the functions of the mTOR pathway. Rapamycin is reported to block protein synthesis, arrest cell cycle progression, and inhibit IL-2 signal transduction mechanisms.