Learn More
TARGETMOL CHEMICALS INC Rapamycin 200MG
SDP
Catalog No. 502875799
Encompass_Preferred
Also available in 1 g, 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Rapamycin (AY 22989) is a natural product of macrolides, an mTOR inhibitor with specificity (HEK293 cells IC50=0.1 nM). Rapamycin has immunosuppressive activity and induces autophagy. Purity 99.03%
Catalog No. 50-287-5799 Supplier TARGETMOL CHEMICALS INC Supplier No. T1537200MG
Also available in 1 g, 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Rapamycin (AY 22989) is a natural product of macrolides, an mTOR inhibitor with specificity (HEK293 cells IC50=0.1 nM). Rapamycin has immunosuppressive activity and induces autophagy. Purity 99.03%