Learn More
Enzo Life Sciences Rapamycin, (25mg), CAS Number: 53123-88-9
SDP
Supplier:  Enzo Life Sciences BMLA2750025
Encompass_Preferred
Immunosuppressant. Alternative Name: Sirolimus. AY-22,989. RAPA. Rapamune. Formula: C51H79NO13. MW: 914.2. CAS Number: 53123-88-9. MI: 14: 8114. Purity: ≥98% (HPLC). Appearance: Off-white or yellow solid. Solubility: Soluble in chloroform, DMSO (25mg/ml) or methanol. Use/Stability: Store, as supplied, at -20°C for up to 1 year. Store solutions at -20°C for up to 3 months. Long Term Storage: -20°C
Catalog No. NC1050897