Learn More
AdipoGen Rapamycin (5 mg)
SDP
Catalog No. 501687269
Encompass_Preferred
Rapamycin (5 mg), Adipogen Life Sciences. Chemical. CAS: 53123-88-9. Formula: C51H79NO13. MW: 914.2. Isolated from Streptomyces hygroscopicus. Antibiotic. Antibacterial and antifungal properties. Forms a complex with FKBP12 and inhibits the mammalian target of rapamycin (mTOR). Inhibits the response to interleukin-2 (IL-2), and thereby blocks activation of T and B cells. Potent immunosuppressant used as an alternative to calcineurin inhibitors. Restricts the proliferation of smooth-muscle cells by blocking cell cycle progression at the G1/S transition. Anti-proliferative. Antitumor compound. Apoptosis enhancer. Activator of autophagy both in vitro and in vivo. Anti-HIV and anti-aging compound. Neuroprotective. Suppressor for self-renewal and vascular differentiation potential in hemangioma stem cells.
Catalog No. 50-168-7269 Supplier AdipoGen Supplier No. AGCN20025M005
Rapamycin (5 mg), Adipogen Life Sciences. Chemical. CAS: 53123-88-9. Formula: C51H79NO13. MW: 914.2. Isolated from Streptomyces hygroscopicus. Antibiotic. Antibacterial and antifungal properties. Forms a complex with FKBP12 and inhibits the mammalian target of rapamycin (mTOR). Inhibits the response to interleukin-2 (IL-2), and thereby blocks activation of T and B cells. Potent immunosuppressant used as an alternative to calcineurin inhibitors. Restricts the proliferation of smooth-muscle cells by blocking cell cycle progression at the G1/S transition. Anti-proliferative. Antitumor compound. Apoptosis enhancer. Activator of autophagy both in vitro and in vivo. Anti-HIV and anti-aging compound. Neuroprotective. Suppressor for self-renewal and vascular differentiation potential in hemangioma stem cells.