Learn More
Alkali Scientific Rapamycin, 5 Mg
SDP
Catalog No. 5000661907
Encompass_Preferred
Rapamycin (5mg) is a potent cell growth inhibitor used extensively in research to study cell signaling and growth regulation. It targets and inhibits mTOR (mechanistic target of rapamycin), a key regulator of cell metabolism, growth, and proliferation. By suppressing mTOR activity, Rapamycin effectively halts cell cycle progression, making it valuable in cancer research, immunology, and aging studies. Additionally, Rapamycin's immunosuppressive properties are utilized in clinical applications to prevent organ rejection in transplant patients. In laboratory settings, it is used to study the effects of mTOR inhibition on cellular responses to stress and nutrient availability.
Catalog No. 50-006-61907 Supplier Alkali Scientific Supplier No. RAP002CP
Rapamycin (5mg) is a potent cell growth inhibitor used extensively in research to study cell signaling and growth regulation. It targets and inhibits mTOR (mechanistic target of rapamycin), a key regulator of cell metabolism, growth, and proliferation. By suppressing mTOR activity, Rapamycin effectively halts cell cycle progression, making it valuable in cancer research, immunology, and aging studies. Additionally, Rapamycin's immunosuppressive properties are utilized in clinical applications to prevent organ rejection in transplant patients. In laboratory settings, it is used to study the effects of mTOR inhibition on cellular responses to stress and nutrient availability.