Learn More
Medchemexpress LLC Rapamycin | 53123-88-9 | 99.9% | 914.17 | 2 G
SDP
Catalog No. 5000365678
Encompass_Preferred
Rapamycin (Sirolimus) is a potent and specific mTOR inhibitor. It binds to FKBP12 and acts as an allosteric inhibitor of mTORC1. Rapamycin is also known as an autophagy activator and an immunosuppressant, and is blood-brain barrier transmissible.
  • Potent and specific mTOR inhibitor
  • Binds to FKBP12
  • Allosteric inhibitor of mTORC1
  • Autophagy activator
  • Immunosuppressant
  • Blood-brain barrier transmissible
Catalog No. 50-003-65678 Supplier Medchemexpress LLC Supplier No. 2GHY10219
Rapamycin (Sirolimus) is a potent and specific mTOR inhibitor. It binds to FKBP12 and acts as an allosteric inhibitor of mTORC1. Rapamycin is also known as an autophagy activator and an immunosuppressant, and is blood-brain barrier transmissible.
  • Potent and specific mTOR inhibitor
  • Binds to FKBP12
  • Allosteric inhibitor of mTORC1
  • Autophagy activator
  • Immunosuppressant
  • Blood-brain barrier transmissible