Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rapamycin | 53123-88-9 | MFCD00867594 | 99.0% | 914.2 g/mol | C51H79NO13 | 100 MG
SDP

Catalog No. 5000246944
Encompass_Preferred

Rapamycin (sirolimus) analytical standard intended for research and analytical applications. Supplied as a 100 mg reference material for assay, calibration, and method development.

  • Analytical standard of rapamycin for research and analytical use.
  • CAS number 53123-88-9.
  • Molecular formula C51H79NO13 and molecular weight 914.2 g/mol.
  • Purity 98.98% (as listed by the manufacturer).
  • Suitable for assay, calibration, and method validation.
  • Synonyms include sirolimus and AY-22989.

Catalog No. 50-002-46944 Supplier Medchemexpress LLC Supplier No. HY10219R100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rapamycin (sirolimus) analytical standard intended for research and analytical applications. Supplied as a 100 mg reference material for assay, calibration, and method development.

  • Analytical standard of rapamycin for research and analytical use.
  • CAS number 53123-88-9.
  • Molecular formula C51H79NO13 and molecular weight 914.2 g/mol.
  • Purity 98.98% (as listed by the manufacturer).
  • Suitable for assay, calibration, and method validation.
  • Synonyms include sirolimus and AY-22989.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.