Learn More
Medchemexpress LLC Rapamycin | 53123-88-9 | MFCD00867594 | 99.9% | 914.17 g/mol | C51H79NO13 | 25 G
SDP
Catalog No. 5000246308
Encompass_Preferred
Rapamycin (sirolimus) is a macrolide small molecule that selectively inhibits mTOR signaling by binding FKBP12. It is used as a research reagent to study cell growth, autophagy, and immunosuppression; the compound is a white to off-white solid with molecular formula C51H79NO13 and molecular weight 914.17 g/mol.
  • Potent mTOR inhibitor that binds FKBP12.
  • High reported purity (99.94% by LCMS).
  • Suitable for research and analytical applications, including autophagy and cell signaling studies.
  • Recommended storage: powder stable at low temperatures; in solution store at -80°C or -20°C depending on duration.
Catalog No. 50-002-46308 Supplier Medchemexpress LLC Supplier No. HY1021925G
Rapamycin (sirolimus) is a macrolide small molecule that selectively inhibits mTOR signaling by binding FKBP12. It is used as a research reagent to study cell growth, autophagy, and immunosuppression; the compound is a white to off-white solid with molecular formula C51H79NO13 and molecular weight 914.17 g/mol.
  • Potent mTOR inhibitor that binds FKBP12.
  • High reported purity (99.94% by LCMS).
  • Suitable for research and analytical applications, including autophagy and cell signaling studies.
  • Recommended storage: powder stable at low temperatures; in solution store at -80°C or -20°C depending on duration.