Learn More
AdipoGen Rapamycin
SDP
Supplier:  AdipoGen AGCN20025M001
Encompass_Preferred
Chemical. CAS 53123-88-9. Formula C51H79NO13. MW 914.2. Isolated from Streptomyces hygroscopicus. Antibiotic. Antibacterial and antifungal properties. Forms a complex with FKBP12 and inhibits the mammalian target of rapamycin mTOR. Inhibits the response to interleukin-2 IL-2, and thereby blocks activation of T and B cells. Potent immunosuppressant used as an alternative to calcineurin inhibitors. Restricts the proliferation of smooth-muscle cells by blocking cell cycle progression at the G1/S transition. Anti-proliferative. Antitumor compound. Apoptosis enhancer. Activator of autophagy both in vitro and in vivo. Anti-HIV and anti-aging compound. Neuroprotective. Suppressor for self-renewal and vascular differentiation potential in hemangioma stem cells.
Catalog No. 50-256-2692