Learn More
Cayman Chemical RapamycIn-d3 1mg
SDP
Catalog No. 502964325
Encompass_Preferred
An internal standard for the quantification of rapamycin by GC- or LC-MS
Catalog No. 50-296-4325 Supplier Cayman Chemical Supplier No. 220931 MG
An internal standard for the quantification of rapamycin by GC- or LC-MS