Learn More
Cayman Chemical RapamycIn-d3 500ug
SDP
Catalog No. 502987400
Encompass_Preferred
An internal standard for the quantification of rapamycin by GC- or LC-MS
Catalog No. 50-298-7400 Supplier Cayman Chemical Supplier No. 22093500 UG
An internal standard for the quantification of rapamycin by GC- or LC-MS