Learn More
Abcam Rapamycin (DMSO solution), immunosuppressant, 5MG-LQD
Catalog No. 502704564
Encompass_Preferred
MW 914.2 Da, Purity >98%. Immunosuppressant and antifungal agent . Complexes with FKBP-12 and inhibits mTOR (mammalian target of rapamycin). In complex with its cellular receptor, the FK506-binding protein (FKBP12), rapamycin binds and inhibits the function of the mammalian target of rapamycin (mTOR).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4564 Supplier Abcam Supplier No. AB1465915MGLQD
MW 914.2 Da, Purity >98%. Immunosuppressant and antifungal agent . Complexes with FKBP-12 and inhibits mTOR (mammalian target of rapamycin). In complex with its cellular receptor, the FK506-binding protein (FKBP12), rapamycin binds and inhibits the function of the mammalian target of rapamycin (mTOR).
The product is subject to the following: Abcam Restricted Use Statement