Learn More
Chemie Tek Rapamycin, 53123-88-9, MFCD00867594, 100mg
SDP
Supplier:  Chemie Tek CT0622100MG
Encompass_Preferred
Mol. Formula: C51H79NO13, >99.5% purity, MW: 914.2, Rapamycin is an immunosuppressant that is used primarily to prevent the rejection of organ and bone marrow transplant. It was first described as a potent inhibitor of interleukin-2 activation of lymphocytes (IC50 = 5 pM). It is now known that rapamycin specifically interacts with the cytosolic FK-binding protein 12 (FKBP12) to form a complex which inhibits the mammalian target of rapamycin (mTOR) pathway by directly binding to mTOR Complex 1 (mTORC1). Rapamycin and other inhibitors of mTORC1 signaling show potential in treating cancer, adipogenesis, diabetes, tuberous sclerosis, and cardiovascular disease.
Catalog No. NC1640568