Learn More
Medchemexpress LLC Rapamycin (sirolimus) | 53123-88-9 | MFCD00867594 | 914.17 g·mol⁻¹ | C51H79NO13 | 100 MG
SDP
Catalog No. 5000245827
Encompass_Preferred
Rapamycin (sirolimus) (GMP) is a GMP-grade macrolide that binds FKBP12 and inhibits mTOR, used in research to modulate cell growth, autophagy, and signaling through the PI3K/Akt/mTOR pathway. It is intended for in vitro and preclinical applications where GMP-like material and batch consistency are required.
  • GMP-grade material suitable for research and preclinical use.
  • Targets mTOR via FKBP12 binding for pathway inhibition.
  • Molecular formula C51H79NO13 and molecular weight 914.17 g·mol⁻¹.
  • Provided as a solid for formulation or experimental preparation.
  • Useful for studies of cell proliferation, autophagy, and signaling modulation.
Catalog No. 50-002-45827 Supplier Medchemexpress LLC Supplier No. HY10219G100MG
Add to Cart
Add to Cart
Request a Quote
Rapamycin (sirolimus) (GMP) is a GMP-grade macrolide that binds FKBP12 and inhibits mTOR, used in research to modulate cell growth, autophagy, and signaling through the PI3K/Akt/mTOR pathway. It is intended for in vitro and preclinical applications where GMP-like material and batch consistency are required.
  • GMP-grade material suitable for research and preclinical use.
  • Targets mTOR via FKBP12 binding for pathway inhibition.
  • Molecular formula C51H79NO13 and molecular weight 914.17 g·mol⁻¹.
  • Provided as a solid for formulation or experimental preparation.
  • Useful for studies of cell proliferation, autophagy, and signaling modulation.