Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rapamycin (sirolimus) | 53123-88-9 | MFCD00867594 | 99.9% | 914.17 g/mol | C51H79NO13 | 50 G
SDP

Catalog No. 5000245887
Encompass_Preferred

Rapamycin (sirolimus) is a macrolide lactone that binds FKBP12 and acts as a potent, selective allosteric inhibitor of mTORC1. It is used in research to modulate autophagy, study mTOR signaling, and as a biochemical standard in cellular and molecular assays.

  • High purity: 99.9% (HPLC)
  • Molecular weight: 914.17 g/mol
  • Chemical formula: C51H79NO13
  • Function: mTOR inhibitor and autophagy activator for cell-based studies
  • Form: supplied as a solid suitable for dissolution and formulation

Catalog No. 50-002-45887 Supplier Medchemexpress LLC Supplier No. HY1021950G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Rapamycin (sirolimus) is a macrolide lactone that binds FKBP12 and acts as a potent, selective allosteric inhibitor of mTORC1. It is used in research to modulate autophagy, study mTOR signaling, and as a biochemical standard in cellular and molecular assays.

  • High purity: 99.9% (HPLC)
  • Molecular weight: 914.17 g/mol
  • Chemical formula: C51H79NO13
  • Function: mTOR inhibitor and autophagy activator for cell-based studies
  • Form: supplied as a solid suitable for dissolution and formulation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.