Learn More
Medchemexpress LLC Rapamycin (sirolimus) | 53123-88-9 | MFCD00867594 | 99.9% | 914.2 g/mol | C51H79NO13 | 5 G
SDP
Catalog No. 5000245888
Encompass_Preferred
Rapamycin (sirolimus) is a macrolide compound and potent mTOR inhibitor supplied as a research-grade solid reagent for laboratory use. It modulates autophagy and immune responses, and is provided with high purity for experimental applications.
  • Macrolide mTOR inhibitor used for research applications.
  • High purity solid suitable for analytical and biological studies.
  • CAS number 53123-88-9 for precise identification.
  • Available in solid quantities including 5 G.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-45888 Supplier Medchemexpress LLC Supplier No. HY102195G
Rapamycin (sirolimus) is a macrolide compound and potent mTOR inhibitor supplied as a research-grade solid reagent for laboratory use. It modulates autophagy and immune responses, and is provided with high purity for experimental applications.
  • Macrolide mTOR inhibitor used for research applications.
  • High purity solid suitable for analytical and biological studies.
  • CAS number 53123-88-9 for precise identification.
  • Available in solid quantities including 5 G.
  • For research use only; not for human or clinical use.