Learn More
Apexbio Technology LLC mTOR inhibitor, antifungal and immunosuppresant compound 53123-88-9. MFCD00867594
SDP
Catalog No. 501011595
Encompass_Preferred
Rapamycin was used as a kind of original antifungal antibiotic, which is produced by Streptomyces hygroscopicus. Now it has been used in the prevention of transplant rejection because of its immunosuppressive effect. It also exhibits activity against several transplantable tumors and slightly activity to inactive against leukemias. The immunosuppressive effect of Rapamycin is exerted by inhibiting the activation and proliferation of T cells. Rapamycin binds to FK-binding protein 12 (FKBP12) and forms the rapamycin-FKBP12 complex, which regulates an enzyme that plays an important role in the progression of the cell cycle. Other sizes are available. Please inqury us for quote.
Catalog No. 50-101-1595 Supplier Apexbio Technology LLC Supplier No. A81675
Rapamycin was used as a kind of original antifungal antibiotic, which is produced by Streptomyces hygroscopicus. Now it has been used in the prevention of transplant rejection because of its immunosuppressive effect. It also exhibits activity against several transplantable tumors and slightly activity to inactive against leukemias. The immunosuppressive effect of Rapamycin is exerted by inhibiting the activation and proliferation of T cells. Rapamycin binds to FK-binding protein 12 (FKBP12) and forms the rapamycin-FKBP12 complex, which regulates an enzyme that plays an important role in the progression of the cell cycle. Other sizes are available. Please inqury us for quote.