Learn More
Medchemexpress LLC Rapamycin (standard) | 53123-88-9 | MFCD00867594 | 99.0% | 914.17 | C51H79NO13 | 5 MG
SDP
Catalog No. 5000246549
Encompass_Preferred
Rapamycin (sirolimus) analytical standard supplied as a 5 mg reference material for research and analytical assays. It is intended for use as a reference in qualitative and quantitative methods, including HPLC, GC, and MS, and represents the active macrolide mTOR inhibitor.
  • Reference standard for HPLC, GC, and MS.
  • Supplied as a 5 mg analytical sample.
  • High purity for accurate quantitation and method development.
  • Intended for research and analytical applications only.
  • Stable when stored according to supplier recommendations.
Catalog No. 50-002-46549 Supplier Medchemexpress LLC Supplier No. HY10219R5MG
Rapamycin (sirolimus) analytical standard supplied as a 5 mg reference material for research and analytical assays. It is intended for use as a reference in qualitative and quantitative methods, including HPLC, GC, and MS, and represents the active macrolide mTOR inhibitor.
  • Reference standard for HPLC, GC, and MS.
  • Supplied as a 5 mg analytical sample.
  • High purity for accurate quantitation and method development.
  • Intended for research and analytical applications only.
  • Stable when stored according to supplier recommendations.