Learn More
Medchemexpress LLC Rapastinel trifluoroacetate | 1435786-04-1 | 98.0% | 527.49 | 1 ML
SDP
Catalog No. 5000393622
Encompass_Preferred
Rapastinel Trifluoroacetate is an NMDA receptor modulator with glycine-site partial agonist properties, showing potential for major depressive disorder treatment.
  • Modulates NMDA receptors as a glycine-site partial agonist
  • Demonstrates antidepressant-like effects in preclinical models
  • Exhibits anxiolytic properties in open field tests
  • Undergoing clinical development for major depressive disorder
  • Available in a 10 mM solution for research use
Catalog No. 50-003-93622 Supplier Medchemexpress LLC Supplier No. HY16728B10MM/1ML
Rapastinel Trifluoroacetate is an NMDA receptor modulator with glycine-site partial agonist properties, showing potential for major depressive disorder treatment.
  • Modulates NMDA receptors as a glycine-site partial agonist
  • Demonstrates antidepressant-like effects in preclinical models
  • Exhibits anxiolytic properties in open field tests
  • Undergoing clinical development for major depressive disorder
  • Available in a 10 mM solution for research use