Learn More
Medchemexpress LLC Rapastinel trifluoroacetate | 1435786-04-1 | MFCD20527320 | 98.0% | 527.49 | C20H32F3N5O8 | 10 MG
SDP
Catalog No. 5000289277
Encompass_Preferred
Rapastinel trifluoroacetate is a peptide-based NMDA receptor modulator used in preclinical research to investigate antidepressant-related mechanisms and receptor pharmacology. It is supplied as a white to off-white solid with high purity and defined storage recommendations.
  • High purity (98.0%) suitable for research applications.
  • Peptide NMDA receptor modulator with glycine-site partial agonist activity.
  • White to off-white solid, convenient for formulation and dosing.
  • Available in small milligram quantities for in vitro and in vivo studies.
  • Recommended storage: sealed, away from moisture; in solvent -80°C (up to 6 months).
Catalog No. 50-002-89277 Supplier Medchemexpress LLC Supplier No. HY16728B10MG
Rapastinel trifluoroacetate is a peptide-based NMDA receptor modulator used in preclinical research to investigate antidepressant-related mechanisms and receptor pharmacology. It is supplied as a white to off-white solid with high purity and defined storage recommendations.
  • High purity (98.0%) suitable for research applications.
  • Peptide NMDA receptor modulator with glycine-site partial agonist activity.
  • White to off-white solid, convenient for formulation and dosing.
  • Available in small milligram quantities for in vitro and in vivo studies.
  • Recommended storage: sealed, away from moisture; in solvent -80°C (up to 6 months).