Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rapeseed Oil, Spectrum™ Chemical
SDP

Catalog No. 18606042 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-042 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-606-042 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. R2010500ML
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

R2010, 8002-13-9

Spectrum™ Chemical Rapeseed Oil

Chemical Identifiers

CAS 8002-13-9

Specifications

CAS Max % 1
Boiling Point 350°C/662°F
Packaging Amber Glass Bottle
Quantity 500 mL
Saponification Value 170-180
Specific Gravity 0.905-0.912
Grade Ungraded
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Rapeseed Oil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.