Learn More
Stanbio™ RaPET RF
SDP
Catalog No. 22022631 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Type:
Reagent
Catalog No. Type
22-022-631 Reagent
1 options
This item is not returnable. View return policy
Catalog No. 22-022-631 Supplier Stanbio™ Supplier No. 1155050
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
Uses Rapid Particle enhanced Technology (RaPET)
  • Test for Rheumatoid Factor
  • No dilution
  • Color-coded dropper caps
  • Superior macroagglutination
Specifications
Type Reagent