Learn More
Aviva Systems Biology Corp RAPGEF1 Antibody - C-terminal region (ARP54753_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54753_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAPGEF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RAPGEF1. Alternate names: C3G, DKFZp781P1719, GRF2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-61301