Learn More
Aviva Systems Biology Corp RAPGEF3 Antibody - middle region (ARP52140_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52140_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAPGEF3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RAPGEF3. Alternate names: CAMP-GEFI, EPAC, EPAC1, MGC21410, bcm910, HSU79275ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-647