Learn More
Medchemexpress LLC Raphin1 acetate | 2242616-04-0 | 99.9% | 291.13 | C10H12Cl2N4O2 | 10MM 1ML
SDP
Catalog No. 5000236864
Encompass_Preferred
Raphin1 acetate is a selective, orally bioavailable inhibitor of the regulatory phosphatase PPP1R15B (R15B). Supplied as a 10 mM solution in DMSO (1 mL) for research use, it is also available in multiple solid sizes. The compound has molecular formula C10H12Cl2N4O2, molecular weight 291.13, CAS 2242616-04-0, and a listed purity of 99.93%.
  • Selective inhibitor of PPP1R15B phosphatase.
  • Ready-to-use 10 mM solution in DMSO for rapid assay setup.
  • High listed purity (99.93%) minimizes contaminants.
  • Suitable for biochemical and cellular research applications.
  • Available in solid forms for custom formulation and dosing.
  • Molecular weight 291.13 supports accurate molarity calculations.
Catalog No. 50-002-36864 Supplier Medchemexpress LLC Supplier No. HY123960A10MM 1ML
Raphin1 acetate is a selective, orally bioavailable inhibitor of the regulatory phosphatase PPP1R15B (R15B). Supplied as a 10 mM solution in DMSO (1 mL) for research use, it is also available in multiple solid sizes. The compound has molecular formula C10H12Cl2N4O2, molecular weight 291.13, CAS 2242616-04-0, and a listed purity of 99.93%.
  • Selective inhibitor of PPP1R15B phosphatase.
  • Ready-to-use 10 mM solution in DMSO for rapid assay setup.
  • High listed purity (99.93%) minimizes contaminants.
  • Suitable for biochemical and cellular research applications.
  • Available in solid forms for custom formulation and dosing.
  • Molecular weight 291.13 supports accurate molarity calculations.