Learn More
Medchemexpress LLC Raphin1 acetate | 2242616-04-0 | 99.9% | 291.13 | C10H12Cl2N4O2 | 25MG
SDP
Catalog No. 5000236443
Encompass_Preferred
Raphin1 acetate is an orally bioavailable, selective small-molecule inhibitor of the regulatory phosphatase PPP1R15B (R15B). It binds the R15B-PP1c holophosphatase with high affinity (Kd = 33 nM) and shows approximately 30-fold selectivity over R15A-PP1c. Supplied as a white to off-white solid, it is intended for biochemical and cellular research applications.
  • Selective inhibitor of regulatory phosphatase PPP1R15B.
  • High-affinity binding to R15B-PP1c (Kd = 33 nM).
  • About 30-fold selectivity versus R15A-PP1c.
  • Available in multiple small-scale packaging options, including 25 mg.
  • High reported purity (≈99.9%) suitable for assays.
  • Solid form simplifies storage and handling.
Catalog No. 50-002-36443 Supplier Medchemexpress LLC Supplier No. HY123960A25MG
Raphin1 acetate is an orally bioavailable, selective small-molecule inhibitor of the regulatory phosphatase PPP1R15B (R15B). It binds the R15B-PP1c holophosphatase with high affinity (Kd = 33 nM) and shows approximately 30-fold selectivity over R15A-PP1c. Supplied as a white to off-white solid, it is intended for biochemical and cellular research applications.
  • Selective inhibitor of regulatory phosphatase PPP1R15B.
  • High-affinity binding to R15B-PP1c (Kd = 33 nM).
  • About 30-fold selectivity versus R15A-PP1c.
  • Available in multiple small-scale packaging options, including 25 mg.
  • High reported purity (≈99.9%) suitable for assays.
  • Solid form simplifies storage and handling.