Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Raphin1 acetate | 2242616-04-0 | 99.9% | 291.13 | C10H12Cl2N4O2 | 50MG
SDP

Catalog No. 5000238362
Encompass_Preferred

Raphin1 acetate is a small-molecule selective inhibitor of the regulatory phosphatase PPP1R15B (R15B) intended for research use. It is orally bioavailable, crosses the blood-brain barrier, and has been used in preclinical studies to reduce molecular and organismal deficits in models of protein misfolding. The compound is supplied as a solid, suitable for laboratory pharmacological and biochemical research.

  • Selective inhibitor of PPP1R15B (R15B).
  • Orally bioavailable and crosses the blood-brain barrier.
  • High purity (99.9%).
  • Molecular weight 291.13; formula C10H12Cl2N4O2.
  • Supplied as a 50 mg solid for research applications.

Catalog No. 50-002-38362 Supplier Medchemexpress LLC Supplier No. HY123960A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Raphin1 acetate is a small-molecule selective inhibitor of the regulatory phosphatase PPP1R15B (R15B) intended for research use. It is orally bioavailable, crosses the blood-brain barrier, and has been used in preclinical studies to reduce molecular and organismal deficits in models of protein misfolding. The compound is supplied as a solid, suitable for laboratory pharmacological and biochemical research.

  • Selective inhibitor of PPP1R15B (R15B).
  • Orally bioavailable and crosses the blood-brain barrier.
  • High purity (99.9%).
  • Molecular weight 291.13; formula C10H12Cl2N4O2.
  • Supplied as a 50 mg solid for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.