Learn More
Medchemexpress LLC Raphin1 acetate | 2242616-04-0 | 99.9% | 291.13 g/mol | C10H12Cl2N4O2 | 10MG
SDP
Catalog No. 5000237092
Encompass_Preferred
Raphin1 acetate is an orally bioavailable small-molecule inhibitor of the regulatory phosphatase PPP1R15B (R15B). It binds the R15B-PP1c holophosphatase with high affinity (Kd ≈ 33 nM), displays about 30-fold selectivity versus R15A-PP1c, and crosses the blood-brain barrier. The compound has shown reduction of molecular and organismal deficits in mouse models of protein misfolding disease and is supplied for research use in multiple small pack sizes.
  • Selective inhibitor of PPP1R15B regulatory phosphatase.
  • High-affinity binding to R15B-PP1c (Kd ≈ 33 nM).
  • Approximately 30-fold selectivity over R15A-PP1c.
  • Crosses the blood-brain barrier for CNS studies.
  • High-purity research reagent suitable for in vitro and in vivo work.
  • Available in multiple small pack sizes for dose-ranging studies.
Catalog No. 50-002-37092 Supplier Medchemexpress LLC Supplier No. HY123960A10MG
Raphin1 acetate is an orally bioavailable small-molecule inhibitor of the regulatory phosphatase PPP1R15B (R15B). It binds the R15B-PP1c holophosphatase with high affinity (Kd ≈ 33 nM), displays about 30-fold selectivity versus R15A-PP1c, and crosses the blood-brain barrier. The compound has shown reduction of molecular and organismal deficits in mouse models of protein misfolding disease and is supplied for research use in multiple small pack sizes.
  • Selective inhibitor of PPP1R15B regulatory phosphatase.
  • High-affinity binding to R15B-PP1c (Kd ≈ 33 nM).
  • Approximately 30-fold selectivity over R15A-PP1c.
  • Crosses the blood-brain barrier for CNS studies.
  • High-purity research reagent suitable for in vitro and in vivo work.
  • Available in multiple small pack sizes for dose-ranging studies.