Learn More
Medchemexpress LLC Raphin1 acetate | 2242616-04-0 | MFCD32201062 | 99.9% | 291.13 g/mol | C10H12Cl2N4O2 | 100MG
SDP
Catalog No. 5000236057
Encompass_Preferred
Raphin1 acetate is a selective, orally bioavailable inhibitor of the regulatory phosphatase PPP1R15B (R15B). It binds the R15B-PP1c holophosphatase with Kd ≈ 33 nM and shows about 30-fold selectivity over R15A-PP1c. Supplied as a high-purity research reagent, it is used to modulate phosphatase activity in studies of proteostasis and protein folding disease models.
  • Selective inhibition of PPP1R15B with strong R15B-PP1c binding.
  • Orally bioavailable, suitable for in vivo research.
  • High chemical purity for reproducible results.
  • Available in multiple pack sizes for experimental flexibility.
  • Applicable to studies of proteostasis and protein-folding disorders.
Catalog No. 50-002-36057 Supplier Medchemexpress LLC Supplier No. HY123960A100MG
Raphin1 acetate is a selective, orally bioavailable inhibitor of the regulatory phosphatase PPP1R15B (R15B). It binds the R15B-PP1c holophosphatase with Kd ≈ 33 nM and shows about 30-fold selectivity over R15A-PP1c. Supplied as a high-purity research reagent, it is used to modulate phosphatase activity in studies of proteostasis and protein folding disease models.
  • Selective inhibition of PPP1R15B with strong R15B-PP1c binding.
  • Orally bioavailable, suitable for in vivo research.
  • High chemical purity for reproducible results.
  • Available in multiple pack sizes for experimental flexibility.
  • Applicable to studies of proteostasis and protein-folding disorders.