Learn More
Cederlane RapiClear 1.52, 10 mL, refractive index 1.52 nD, one-step tissue clearing, water-soluble
SDP
Catalog No. NC1633209
Encompass
RapiClear 1.52 is a ready-to-use, water-soluble tissue clearing reagent designed to enhance the visualization of biological specimens by facilitating swift transparency of various tissues. This product aids in the construction of detailed 3D images while minimizing sample deformation and reducing opacity. With a refractive index of 1.52 nD, it optimizes imaging clarity by closely matching that of glass and immersion oil.
  • Ready-to-use for immediate application
  • Water-soluble composition for easy use
  • One-step tissue clearing for efficiency
  • Transparent effect is reversible
  • Visualization depth up to 1-5 mm
  • Limited sample deformation during the clearing process
  • Compatible with various tissue types including whole mouse organs and plant tissues
  • Recommended pre-warming to 37℃ before mounting
  • Stability lasts at least 1 year when stored correctly
Catalog No. NC1633209 Supplier Cederlane Supplier No. RC152001
RapiClear 1.52 is a ready-to-use, water-soluble tissue clearing reagent designed to enhance the visualization of biological specimens by facilitating swift transparency of various tissues. This product aids in the construction of detailed 3D images while minimizing sample deformation and reducing opacity. With a refractive index of 1.52 nD, it optimizes imaging clarity by closely matching that of glass and immersion oil.
  • Ready-to-use for immediate application
  • Water-soluble composition for easy use
  • One-step tissue clearing for efficiency
  • Transparent effect is reversible
  • Visualization depth up to 1-5 mm
  • Limited sample deformation during the clearing process
  • Compatible with various tissue types including whole mouse organs and plant tissues
  • Recommended pre-warming to 37℃ before mounting
  • Stability lasts at least 1 year when stored correctly