Learn More
Cell Signaling Technology Rapid-Act T Cell Activation Kit, Human, Anti-CD3/CD28, 50 Tests
SDP
Catalog No. NC2888739
Encompass
The Rapid-Act T Cell Activation Kit (Human, Anti-CD3/CD28) provides the necessary reagents for easy, single-step induction of human T cell activation and proliferation. The included Synthetic Immune Synapse particles pre-cluster CD3 and CD28 antibodies for thorough T cell activation while minimizing cell clumping. The kit supports accurate time course studies for short-term stimulation, as well as longer-term T cell proliferation and exhaustion assays, with no modification or removal necessary prior to downstream flow cytometry.
  • Enables easy, single-step induction of T cell activation and proliferation
  • Includes Synthetic Immune Synapse particles to pre-cluster antibodies
  • Minimizes cell clumping
  • Supports both short-term stimulation and long-term assays
  • Compatible with flow cytometry analysis without reagent removal
  • Suitable for 50 assays
  • Species reactivity: Human
Catalog No. NC2888739 Supplier Cell Signaling Technology Supplier No. 88179S
The Rapid-Act T Cell Activation Kit (Human, Anti-CD3/CD28) provides the necessary reagents for easy, single-step induction of human T cell activation and proliferation. The included Synthetic Immune Synapse particles pre-cluster CD3 and CD28 antibodies for thorough T cell activation while minimizing cell clumping. The kit supports accurate time course studies for short-term stimulation, as well as longer-term T cell proliferation and exhaustion assays, with no modification or removal necessary prior to downstream flow cytometry.
  • Enables easy, single-step induction of T cell activation and proliferation
  • Includes Synthetic Immune Synapse particles to pre-cluster antibodies
  • Minimizes cell clumping
  • Supports both short-term stimulation and long-term assays
  • Compatible with flow cytometry analysis without reagent removal
  • Suitable for 50 assays
  • Species reactivity: Human