Learn More
Germaine™ Laboratories Rapid Check™ Respiratory Syncytial Virus RSV Antigen Test
Description
- Rapid, qualitative detection of respiratory syncytial virus fusion protein in nasopharyngeal swabs or nasal aspirate specimens in children less than 6 years and adults over 60.
- Intended as an aid in the rapid laboratory diagnosis of acute respiratory syncytial virus infection in patients with such symptoms.
- 510(k) Cleared and CLIA Waived for Prescription Use Detection of fusion protein of respiratory syncytial virus Nasopharyngeal swab and nasal aspirate specimen
- Each kit includes:
- 20 Individually packaged test devices
- Nasopharyngeal Swabs with each kit
- 1 R1 (Extraction Reagent Solution) 2 mL
- 20 Extraction Tubes
- 40 Disposable Pipettes
- 1 PC (Positive Control Reagent)
- 1.0 mL 1 NC (Negative Control Reagent)
- 1 Instructions for Use
- Premier Test Performance: Positive % Agreement: 93. 9% Negative % Agreement: 97.7%
Specifications
Specifications
| Control Sets | Positive and Negative |
| CPT Code | 87807 |
| CE Marker | No |
| Clia Complexity | Waived |
| Format | Test Kit |
| Sample Type | Nasopharyngeal Swabs, Nasal Aspirates |
| Type | RSV Rapid Test |
| For Use With (Application) | Testing children less than 6 years old and adults over 60 years. |
| Includes | 20 Individually packaged test devices, Nasopharyngeal Swabs with each kit, 1 R1 (Extraction Reagent Solution) 2 mL, 20 Extraction Tubes, 40 Disposable Pipettes, 1 PC (Positive Control Reagent) 1.0 mL, 1 NC (Negative Control Reagent) 1.0 mL, 1 Instructions for Use |
| Quantity | 20 tests/kit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.