Learn More
Thomson Instrument Company Rapid Clear Cap 3000, High-Speed Clarification of 2L-4L of Cell Culture, Sterile
SDP
Catalog No. NC1299811
Encompass_Preferred
Thomson’s Rapid Clear Cap is a breakthrough clarification solution that streamlines downstream processing by eliminating centrifugation and simplifying workflows.
•Clarify 3L of culture in under 30 minutes—no centrifugation required!
•0.2 μm final pore size depth filtration eliminates multiple filtration steps in one streamlined setup
•Clarifies low or high-density cultures of CHO stable, CHO transient, HEK293, hybridoma, and other mammalian cell lines
•Eliminates transfer steps—cap screws directly onto Optimum Growth Flasks
•Secondary port connects to fresh flasks or storage vessels via Luer lock
•Enables walk-away convenience and reduces endotoxin exposure for safer workflows
•Reduces single-use consumables by up to 90%, promoting greener lab operations
•Compatible with 2.8L and 5L Optimum Growth Flasks
•Made in the USA and cleanroom-ready (individually packaged and sterilized)
Catalog No. NC1299811 Supplier Thomson Instrument Company Supplier No. 788116
Thomson’s Rapid Clear Cap is a breakthrough clarification solution that streamlines downstream processing by eliminating centrifugation and simplifying workflows.
•Clarify 3L of culture in under 30 minutes—no centrifugation required!
•0.2 μm final pore size depth filtration eliminates multiple filtration steps in one streamlined setup
•Clarifies low or high-density cultures of CHO stable, CHO transient, HEK293, hybridoma, and other mammalian cell lines
•Eliminates transfer steps—cap screws directly onto Optimum Growth Flasks
•Secondary port connects to fresh flasks or storage vessels via Luer lock
•Enables walk-away convenience and reduces endotoxin exposure for safer workflows
•Reduces single-use consumables by up to 90%, promoting greener lab operations
•Compatible with 2.8L and 5L Optimum Growth Flasks
•Made in the USA and cleanroom-ready (individually packaged and sterilized)