Learn More
Research Products International Corp Rapid Commassie Stain, 100 Milliliters
SDP
Catalog No. 50212965
Encompass_Preferred
This unique Coomassie Blue based formulation features a special dye that is absorbed by protein bands instead of the gel matrix. This eliminates the need for destaining and offers enhanced sensitivity and sharper resolution of bands. Protein bands can be detected within 10 minutes. Maximum intensification may require incubation up to 40 minutes. Offers greater convenience and sensitivity than conventional stains. Compatible with silver staining. Rapid Coomassie stain uses a simple two-step procedure. Stain is designed to detect as little as 7.9ng of protein within one hour of electrophoresis.
Supplied as 20X concentrated solution.
Shelf life is approximately 18 months at room temperature.
Catalog No. 50-212-965 Supplier Research Products International Corp Supplier No. RCS50
This unique Coomassie Blue based formulation features a special dye that is absorbed by protein bands instead of the gel matrix. This eliminates the need for destaining and offers enhanced sensitivity and sharper resolution of bands. Protein bands can be detected within 10 minutes. Maximum intensification may require incubation up to 40 minutes. Offers greater convenience and sensitivity than conventional stains. Compatible with silver staining. Rapid Coomassie stain uses a simple two-step procedure. Stain is designed to detect as little as 7.9ng of protein within one hour of electrophoresis.
Supplied as 20X concentrated solution.
Shelf life is approximately 18 months at room temperature.