Learn More
Bioworld Rapid Coomassie Stain, 200 mL
Catalog No. 50198715
Encompass_Preferred
Item# 21470008This stain is fast and sensitive in staining proteins in acrylimide gels. Its features include: a) completion within 30-60 minutes, b) unnecessary destaining, c) being highly sensitive, it detects as little as 30 ng of protein, d) yields sharper bands than those stained with conventional Coomassie Blue, and e) gels can be restained with Silver. Solution provided as 2x concentrate.
Catalog No. 50-198-715 Supplier Bioworld Supplier No. 214700082
Item# 21470008This stain is fast and sensitive in staining proteins in acrylimide gels. Its features include: a) completion within 30-60 minutes, b) unnecessary destaining, c) being highly sensitive, it detects as little as 30 ng of protein, d) yields sharper bands than those stained with conventional Coomassie Blue, and e) gels can be restained with Silver. Solution provided as 2x concentrate.