Learn More
Electron Microscopy Sciences EMS Rapid-Core Cutting Mat
SDP
Supplier:  Electron Microscopy Sciences 6903990
Encompass_Preferred
Five layer composite cutting mat will let you cut cleanly through an object and into the mat. Great for protecting both cutting tip and working surface. Ensures the cored sample comes out completely without burrs and bits. These cutting mats are reversible, inert, self-healing and can be sterilized with ethanol. 1.5 mm thick.
Catalog No. 50-192-7741