Learn More
Electron Microscopy Sciences EMS-Core Sampling Tool | Type: 3mm | ID: 3mm | OD: 3.4mm | Wall Thickness: 0.20
SDP
Catalog No. 501927737
Encompass_Preferred
EMS-Core is an all new revolutionary multi-purpose sampling tool. It is designed with retractable cutting cannulla for easy operation. Efficient, safe, fast and precise, the EMS-Core consists of a razor sharp stainless steel cutting tip, designed to cut, retrieve and store cored samples from source materials such as skin, tissue, gels, paper, cloth, leaves, paint chips, films or other thin, soft substrates. The EMS-Core is well known and recommended as the special DNA sampling tool and is ideal for forensic and laboratory sampling purposes. Each EMS-Core is individually packaged and ethylene oxide sterilized.
Catalog No. 50-192-7737 Supplier Electron Microscopy Sciences Supplier No. 6903930
EMS-Core is an all new revolutionary multi-purpose sampling tool. It is designed with retractable cutting cannulla for easy operation. Efficient, safe, fast and precise, the EMS-Core consists of a razor sharp stainless steel cutting tip, designed to cut, retrieve and store cored samples from source materials such as skin, tissue, gels, paper, cloth, leaves, paint chips, films or other thin, soft substrates. The EMS-Core is well known and recommended as the special DNA sampling tool and is ideal for forensic and laboratory sampling purposes. Each EMS-Core is individually packaged and ethylene oxide sterilized.