Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rapid Decalcifier Solution, OSTEOMOLL™, MilliporeSigma™

Catalog No. M1017362500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1017362500 2.5 L
M1017361000 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1017362500 Supplier MilliporeSigma Supplier No. 1017362500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For histology
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Chemical Name or Material Rapid Decalcifier Solution
CAS 7647-01-0, 50-00-0, 67-56-1
Odor Stinging
Packaging Plastic bottle
Color Blue
Quantity 2.5 L
UN Number UN 1789
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.