Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Rapid-DNA Blood/Cell/Tissue Kit

Catalog No. MP116520005 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Matrix:
Without Matrix
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Matrix
MP116520005 Without Matrix
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MP116520005 Supplier MP Biomedicals™ Supplier No. 116520005

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapid-DNA Blood/Cell/Tissue Kit is used for rapid isolation of high purity, ready-to-use genomic DNA.

The Rapid-DNA Blood/Cell/Tissue Kit is a fast and easy method to obtain high quality genomic DNA from whole blood, cultured cells, buffy coats, tissues and other body fluids. The easy to use, rapid DNA extraction protocols can process multiple types of samples in a single kit and is ideal for the extraction of pure, ready-to-use, high molecular weight genomic DNA in a minimal amount of time. Genomic DNA extracted with this kit can be used with all downstream applications such as PCR, qPCR, Next Generation sequencing, restriction digests and cloning.

MP Biomedical provides the fully equipped Rapid-DNA extraction kit to isolate genomic DNA from any sample type of blood, cells, and tissues at high quality for research analysis and downstream applications.

  • High-Quality DNA Purification
  • Pure genomic DNA Isolation
  • All-in-One Kit for Blood, Cells, and Tissue
  • Ideal for all Downstream Applications (ie. PCR, Restriction Digestion, Sequencing)

In this Rapid-DNA extraction kit, lysing matrix is provided to process a range of tissues and/or organs. Samples are placed into 2.0 mL tubes containing Lysing Matrix M, a 1/4-inch ceramic beads designed to efficiently lyse a variety of tissue samples. Homogenization in a FastPrep instrument with Lysing Matrix M takes place in the presence of lysing buffer that is provided within this kit and validated to protect and solubilize nucleic acids and proteins upon cell lysis.

Depending on the expected yield of genomic DNA, the DNA can also be eluted with a lower or a higher volume of Elution Buffer. The minimum elution volume is 50 μL. If quite large amount of DNA is expected, the volume of elution can be increased (100 to 200 μL). To maximize the final yield, we recommend a second elution step with an equal volume of Elution Buffer.

TRUSTED_SUSTAINABILITY

Specifications

Column Type Silica Spin Filter
Format Kit
Sample Type Whole Blood, Cultured Cells, Buffy Coats, Tissues, Body Fluids, Cell Cultures, Buffy Coat, Tissue, Body Fluids
Elution Volume 50 to 200 μL
For Use With (Application) Blood, cells and tissues to isolate genomic DNA
Matrix Without Matrix
No. of Reactions 50
Nucleic Acid Type Genomic DNA
Product Type DNA Isolation Kit
Shipping Condition Ambient

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.