Learn More
Hudson Surface Technology RAPID ENZYME DIGEESTION SYSTEM
Catalog No. NC3584392
Encompass
  • High efficiency for enzymatic sample preparation
  • Rapid digestion times of 5-10 min
  • Single sample to 96 samples per run capacity
  • Low-frequency microwave heating method
  • ±0.5 °C temperature control accuracy
  • Applications in proteolysis and glycan separation
  • Compatible with multiple enzymes including Trypsin and PNGase F
  • Improves workflow efficiency and sequence coverage
  • 500 mL water volume capacity
  • Cost-effective solution for high-throughput analysis
Catalog No. NC3584392 Supplier Hudson Surface Technology Supplier No. SPM02010001435
Add to Cart
Add to Cart
Request a Quote
  • High efficiency for enzymatic sample preparation
  • Rapid digestion times of 5-10 min
  • Single sample to 96 samples per run capacity
  • Low-frequency microwave heating method
  • ±0.5 °C temperature control accuracy
  • Applications in proteolysis and glycan separation
  • Compatible with multiple enzymes including Trypsin and PNGase F
  • Improves workflow efficiency and sequence coverage
  • 500 mL water volume capacity
  • Cost-effective solution for high-throughput analysis