Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Rapid Gold BCA Protein Assay Kit

Catalog No. p-7111287
Change view
Click to view available options
Quantity:
20 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA53227 20 mL
PIA53226 250 mL
PIA53225 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Store at 4°C
Catalog No. PIA53227 Supplier Thermo Scientific™ Supplier No. A53227
Store at 4°C

Thermo Scientific™ Pierce™ Rapid Gold BCA Protein Assay Kit is two-component, high-precision, detergent-compatible assay optimized to measure (at 480 nm) total protein concentration compared to protein standard curve of known concentrations.

  • A fast, two-component, high-precision, detergent-compatible assay optimized to determine total protein concentration
  • Can be used to assess yields in whole cell lysates and affinity-column fractions, as well as to monitor protein contamination in industrial applications
  • Assay maintains the key characteristics of the traditional BCA assay but allows a fast time and room temperature incubation equal to dye-binding methods
  • Provides comparable accuracy but with a 5 min., room temperature incubation and measured at 480 nm
  • This improvement eliminates the need to wait or expose the samples to elevated temperatures for a fast time to results
  • Exhibits similar protein-to-protein variation as traditional BCA Protein Assay and less protein-to-protein variation than dye-binding protein assay methods (Bradford)
  • Unaffected by typical concentrations of most ionic and non-ionic detergents
  • Linear working range for BSA of 20 to 2000 μg/mL
  • Store in refrigerator at 2°C to 8°C
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 20 to 2000 μg/mL
Content And Storage Store in refrigerator (2–8°C).
Assay Rapid Gold BCA Assay
Detection Method Colorimetric
Product Line Pierce
Product Type Protein Quantitation Assay
Specificity Not Target-Specific
For Use With (Application) Solution-based Detection, Absorbance
For Use With (Equipment) Spectrophotometer, Microplate Reader
Quantity 20 mL
Shipping Condition Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.