Learn More
Fd Neurotechnologies Inc RAPID GOLGISTAIN SOLN C
SDP
Catalog No. NC0486289
Encompass
Fd Rapid Golgistain Kit Is Designed Based On The Principle Of The Methods Described By Ram�n- Moliner, Glaser And Van Der Loos. This Kit Has Not Only Dramatically Improved And Simplified The Golgi-Cox Technique But Has Also Proven To Be Extremely Reliable And Sensitive For Demonstrating Morphological Details Of Neurons And Glia, Especially Dendritic Spines. The Fd Rapid Golgistain Kit Has Been Tested Extensively And Widely Used On The Brains From Several Species Of Animals As Well As On The Specimens Of Postmortem Human Brains. Size For Up To 50 Mouse Brains.
Catalog No. NC0486289 Supplier Fd Neurotechnologies Inc Supplier No. PK401C
Fd Rapid Golgistain Kit Is Designed Based On The Principle Of The Methods Described By Ram�n- Moliner, Glaser And Van Der Loos. This Kit Has Not Only Dramatically Improved And Simplified The Golgi-Cox Technique But Has Also Proven To Be Extremely Reliable And Sensitive For Demonstrating Morphological Details Of Neurons And Glia, Especially Dendritic Spines. The Fd Rapid Golgistain Kit Has Been Tested Extensively And Widely Used On The Brains From Several Species Of Animals As Well As On The Specimens Of Postmortem Human Brains. Size For Up To 50 Mouse Brains.