Learn More
HIMEDIA Rapid HiColiform Broth, M1453, Microbiology Culture Media, Dehydrated Culture Media, Chromogenic Broth, E. coli and Total Coliforms, Water Testing, Powder, 500 g
Catalog No. NC4013689
Encompass_Preferred
Rapid HiColiform Broth detects and confirms Escherichia coli and total coliforms from water in a single step using enzyme-substrate chemistry. A chromogenic substrate cleaved by coliform beta-galactosidase develops a distinct colour, while a fluorogenic substrate cleaved by E. coli beta-glucuronidase fluoresces under UV light, allowing simultaneous read-out. Special peptone and selective sodium lauryl sulphate support sensitivity and specificity. A fast, reliable choice for drinking and environmental water microbiology. Dehydrated powder, store in a cool dry place. Packaged as 500 g.
Catalog No. NC4013689 Supplier HIMEDIA Supplier No. M1453500G
Rapid HiColiform Broth detects and confirms Escherichia coli and total coliforms from water in a single step using enzyme-substrate chemistry. A chromogenic substrate cleaved by coliform beta-galactosidase develops a distinct colour, while a fluorogenic substrate cleaved by E. coli beta-glucuronidase fluoresces under UV light, allowing simultaneous read-out. Special peptone and selective sodium lauryl sulphate support sensitivity and specificity. A fast, reliable choice for drinking and environmental water microbiology. Dehydrated powder, store in a cool dry place. Packaged as 500 g.