Learn More
Diagnostic Biosystems RAPID HISTO-SEALER 10X Rapid Histo-Sealer (10X)™
Catalog No. 502407362
Encompass_Preferred
Rapid Histo-Sealer™ is a preservation solution that acts to rapidly dehydrate tissues mounted on microscope slides in preparation for permanent mounting. This quick-acting dehydrating solution is ideal for rapid immunohistochemistry procedures such as Mohs micrographic surgery. This solution seals the tissue to protect the tissues from drying artifacts to and improve tissue morphology. When applied to the stained section, specimens can be permanently mounted without chromogens fading. Additional benefits of the Rapid Histo-Sealer™ include: saves hands on time when mounting slides, avoids the use of toxic chemicals such as xylene and eliminates accumulation of hazardous waste.\n\nThis product is designed for tissue specimens stained with peroxidase, alkaline phosphatase-based methods using a variety ofchromogens.
Catalog No. 50-240-7362 Supplier Diagnostic Biosystems Supplier No. K093
Rapid Histo-Sealer™ is a preservation solution that acts to rapidly dehydrate tissues mounted on microscope slides in preparation for permanent mounting. This quick-acting dehydrating solution is ideal for rapid immunohistochemistry procedures such as Mohs micrographic surgery. This solution seals the tissue to protect the tissues from drying artifacts to and improve tissue morphology. When applied to the stained section, specimens can be permanently mounted without chromogens fading. Additional benefits of the Rapid Histo-Sealer™ include: saves hands on time when mounting slides, avoids the use of toxic chemicals such as xylene and eliminates accumulation of hazardous waste.\n\nThis product is designed for tissue specimens stained with peroxidase, alkaline phosphatase-based methods using a variety ofchromogens.