Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics Rapid Influenza A/B Control Panel
SDP

Catalog No. 21402644 Shop All Microbiologics Products
Change view
Click to view available options
Form:
Swab
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
21-402-644 Swab
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-402-644 Supplier Microbiologics Supplier No. 8269
Add to Cart
Add to Cart

QC sets and panels, helix elite™ contains 6 positive control foam swabs and 6 negative control foam swabs

  • Positive Control
  • Influenza A Virus (H1N1-2009)
  • Influenza B Virus B/Brisbane
  • Negative Control
  • Blank control swab (Foam swabs with lyophilization buffer)

Specifications

Volume (Metric) 20 μL
Filtered Filtered
Sterility Sterile
Detectable Analytes Influenza A and B
Sample Type Swab
Type Control Panel
Disposable Single-use
Format Rack
Tip Style Extended Length
Form Swab
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.