Learn More
Raybiotech Inc Rapid Mouse Ig Isotyping Array 1
Supplier:  Raybiotech Inc AAMISOG11
Encompass_Preferred
RayBio Rapid Mouse Immunoglobulin Isotyping Array 1. Detects 8 Mouse Ig sub-classes and 2 light chain types. Suitable for serum, plasma, and cell culture supernatants.
Catalog No. NC1957183