Learn More
Cliawaived Inc IDTC Rapid Nicotine Test Cards
SDP
Catalog No. NC1859633
Encompass_Preferred
(40 Tests/Kit)The first ever 3 line lateral flow format. The most sensitive rapid nicotine test available on the US market with ultra-low cutoff of 50 ng/mL.-All results can be read at 5 minutes, but are stable for 60 minutes-Result stability allows for walk-away capability-Test card is hermetically-sealed (leakproof)-Card can be left submerged in specimen throughout the testing process
This item is not returnable. View return policy
Catalog No. NC1859633 Supplier Cliawaived Inc Supplier No. CLIANICOTINE
(40 Tests/Kit)The first ever 3 line lateral flow format. The most sensitive rapid nicotine test available on the US market with ultra-low cutoff of 50 ng/mL.-All results can be read at 5 minutes, but are stable for 60 minutes-Result stability allows for walk-away capability-Test card is hermetically-sealed (leakproof)-Card can be left submerged in specimen throughout the testing process